Assay Details
Target Gene Details
Entrez Gene ID: | 669 |
Gene Name: | bisphosphoglycerate mutase |
Gene Aliases: |
DPGM |
Location: |
Chr.7:134646779-134679816 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BPGM | NM_001293085.1 | NP_001280014.1 | ||
NM_001724.4 | NP_001715.1 | |||
NM_199186.2 | NP_954655.1 | |||
DB047822.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv5958 | Chr.7:134622955 - 134656805 on Build GRCh38 | Insertion | BPGM |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |