Assay Details
Target Gene Details
Entrez Gene ID: | 55033 |
Gene Name: | FK506 binding protein 14 |
Gene Aliases: |
EDSKMH, FKBP22, IPBP12 |
Location: |
Chr.7:30010583-30026801 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FKBP14 | NM_017946.3 | NP_060416.1 | ||
NR_046478.1 | ||||
NR_046479.1 | ||||
AK000738.1 | BAA91351.1 | |||
AK309552.1 | ||||
AY358643.1 | AAQ89006.1 | |||
BC005206.1 | AAH05206.1 | |||
DB498479.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1019527 | Chr.7:29722658 - 30413302 on Build GRCh38 | Gain | SCRN1 WIPF3 LOC105375218 MIR550B1 LOC101928208 ZNRF2 MTURN FKBP14 MIR550A1 LINC01176 PLEKHA8 DKFZP586I1420 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |