Assay Details
Target Gene Details
Entrez Gene ID: | 10165 |
Gene Name: | solute carrier family 25 member 13 |
Gene Aliases: |
ARALAR2, CITRIN, CTLN2 |
Location: |
Chr.7:96120220-96322147 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC25A13 | NM_001160210.1 | NP_001153682.1 | ||
NM_014251.2 | NP_055066.1 | |||
NR_027662.1 | ||||
XM_006715831.3 | XP_006715894.1 | |||
XM_017011663.1 | XP_016867152.1 | |||
XM_017011664.1 | XP_016867153.1 | |||
XM_017011665.1 | XP_016867154.1 | |||
AF118838.1 | AAD38501.1 | |||
AJ496569.1 | CAD43091.1 | |||
AK000766.1 | ||||
AK025227.1 | ||||
AK025779.1 | ||||
AK222864.1 | BAD96584.1 | |||
AK294629.1 | ||||
BC006566.2 | AAH06566.1 | |||
Y17571.1 | CAB62206.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)