Assay Details
Target Gene Details
Entrez Gene ID: | 64921 |
Gene Name: | CAS1 domain containing 1 |
Gene Aliases: |
C7orf12, NBLA04196 |
Location: |
Chr.7:94509858-94633465 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CASD1 | NM_022900.4 | NP_075051.4 | ||
XM_006716092.3 | XP_006716155.1 | |||
XM_006716093.3 | XP_006716156.2 | |||
XM_011516495.2 | XP_011514797.1 | |||
XM_017012546.1 | XP_016868035.1 | |||
XM_017012547.1 | XP_016868036.1 | |||
AF355594.1 | AAL33538.1 | |||
AF397424.2 | AAK97479.2 | |||
AK123866.1 | ||||
AK314329.1 | ||||
BC063284.1 | AAH63284.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1023193 | Chr.7:93602376 - 95828660 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)