Assay Details
Target Gene Details
Entrez Gene ID: | 908 |
Gene Name: | chaperonin containing TCP1 subunit 6A |
Gene Aliases: |
CCT-zeta, CCT-zeta-1, CCT6, Cctz, HTR3, MoDP-2, TCP-1-zeta, TCP20, TCPZ, TTCP20 |
Location: |
Chr.7:56051685-56063989 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCT6A | NM_001009186.1 | NP_001009186.1 | ||
NM_001762.3 | NP_001753.1 | |||
AB063318.1 | BAB61032.1 | |||
AB209728.1 | BAD92965.1 | |||
AF385084.1 | AAK61354.1 | |||
AK297751.1 | ||||
AK298371.1 | ||||
AK313819.1 | ||||
AK316230.1 | ||||
AM295155.1 | ||||
BC106942.1 | AAI06943.1 | |||
BI464886.1 | ||||
BU540578.1 | ||||
L27706.1 | AAA61061.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3613258 | Chr.7:55963379 - 56119128 on Build GRCh38 | Gain | PSPH SNORA15 NUPR2 SUMF2 CCT6A CHCHD2 GBAS PHKG1 |
esv2761334 | Chr.7:55755888 - 56357046 on Build GRCh38 | Gain | PSPH ZNF713 SNORA15 NUPR2 PSPHP1 SEPT14 MRPS17 SUMF2 CCT6A CHCHD2 GBAS PHKG1 |
dgv1201e199 | Chr.7:55674375 - 56426809 on Build GRCh38 | Deletion | ZNF713 FKBP9P1 PSPHP1 MRPS17 SUMF2 LOC101060341 LOC107986800 PSPH LOC650226 SNORA15 NUPR2 SEPT14 CCT6A CHCHD2 GBAS PHKG1 |
esv3576357 | Chr.7:56000465 - 56087692 on Build GRCh38 | Gain | PSPH SNORA15 SUMF2 CCT6A PHKG1 |
esv28230 | Chr.7:55552107 - 56317719 on Build GRCh38 | Gain+Loss | ZNF713 VOPP1 FKBP9P1 PSPHP1 MRPS17 SUMF2 LOC101060341 PSPH SNORA15 NUPR2 SEPT14 CCT6A CHCHD2 GBAS PHKG1 |
esv2734448 | Chr.7:55785310 - 57103280 on Build GRCh38 | Deletion | ZNF713 LOC101928401 LOC100240728 MRPS17 SUMF2 LOC100130849 LOC107986800 MIR4283-1 PSPH LOC650226 SNORA15 NUPR2 LOC100287294 SEPT14 CCT6A CHCHD2 LOC401357 GBAS PHKG1 DKFZp434L192 |
esv3613248 | Chr.7:55667080 - 56415551 on Build GRCh38 | Gain | ZNF713 FKBP9P1 PSPHP1 MRPS17 SUMF2 LOC101060341 LOC107986800 PSPH SNORA15 NUPR2 SEPT14 CCT6A CHCHD2 GBAS PHKG1 |
esv3613259 | Chr.7:56049407 - 56329524 on Build GRCh38 | Gain | PSPH SNORA15 NUPR2 SUMF2 CCT6A CHCHD2 PHKG1 |
esv2734447 | Chr.7:55664175 - 56422045 on Build GRCh38 | Deletion | ZNF713 FKBP9P1 PSPHP1 MRPS17 SUMF2 LOC101060341 LOC107986800 PSPH SNORA15 NUPR2 SEPT14 CCT6A CHCHD2 GBAS PHKG1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |