Assay Details
Target Gene Details
Entrez Gene ID: | 2770 |
Gene Name: | G protein subunit alpha i1 |
Gene Aliases: |
Gi |
Location: |
Chr.7:80134824-80219409 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GNAI1 | NM_001256414.1 | NP_001243343.1 | ||
NM_002069.5 | NP_002060.4 | |||
AF055013.1 | AAC09361.1 | |||
AF493905.1 | AAM12619.1 | |||
AI878901.1 | ||||
AK292953.1 | ||||
AK304442.1 | ||||
AL049933.1 | CAB43212.2 | |||
BC026326.1 | AAH26326.1 | |||
BT019775.1 | AAV38580.1 | |||
M17219.1 | AAA52581.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |