Assay Details
Target Gene Details
Entrez Gene ID: | 404744 |
Gene Name: | NPSR1 antisense RNA 1 |
Gene Aliases: |
AAA1 |
Location: |
Chr.7:34346512-34834331 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NPSR1-AS1 | NR_033664.1 | |||
NR_033665.1 | ||||
AY312368.1 | AAQ85136.1 | |||
AY312369.1 | AAQ85137.1 | |||
AY312370.1 | AAQ85138.1 | |||
AY321516.1 | AAQ87610.1 | |||
BX649082.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv1080e201 | Chr.7:32621750 - 35008557 on Build GRCh38 | Deletion | LOC100421336 RP9 MIR550B2 LINC00997 NT5C3A DPY19L1 MIR548N NPSR1 BBS9 MIR550A2 ZNRF2P1 FLJ20712 DPY19L1P2 FKBP9 RP9P DPY19L1P1 BMPER KBTBD2 NPSR1-AS1 |
nsv1030687 | Chr.7:34361654 - 34438508 on Build GRCh38 | Loss | NPSR1-AS1 |
esv3612796 | Chr.7:34363413 - 34431285 on Build GRCh38 | Loss | NPSR1-AS1 |
esv3891097 | Chr.7:33915559 - 34483233 on Build GRCh38 | Gain | BMPER NPSR1-AS1 |
esv3612772 | Chr.7:33730657 - 34432276 on Build GRCh38 | Loss | BMPER NPSR1-AS1 |
nsv1024493 | Chr.7:33733357 - 34425137 on Build GRCh38 | Loss | BMPER NPSR1-AS1 |
nsv1135982 | Chr.7:32794601 - 34893593 on Build GRCh38 | Deletion | FLJ20712 DPY19L1P2 LOC100421336 FKBP9 RP9 RP9P BMPER NT5C3A KBTBD2 NPSR1 NPSR1-AS1 BBS9 |
nsv5692 | Chr.7:34359469 - 34404413 on Build GRCh38 | Deletion | NPSR1-AS1 |
nsv436907 | Chr.7:32668474 - 34999437 on Build GRCh38 | Insertion | LOC100421336 RP9 MIR550B2 LINC00997 NT5C3A DPY19L1 MIR548N NPSR1 BBS9 MIR550A2 ZNRF2P1 FLJ20712 DPY19L1P2 FKBP9 RP9P DPY19L1P1 BMPER KBTBD2 NPSR1-AS1 |