Assay Details
Target Gene Details
Entrez Gene ID: | 2051 |
Gene Name: | EPH receptor B6 |
Gene Aliases: |
HEP |
Location: |
Chr.7:142854640-142871093 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPHB6 | NM_001280794.2 | NP_001267723.2 | ||
NM_001280795.2 | NP_001267724.2 | |||
NM_004445.5 | NP_004436.4 | |||
NR_104001.2 | ||||
XM_006715881.2 | XP_006715944.1 | |||
XM_011515879.1 | XP_011514181.1 | |||
XM_011515880.1 | XP_011514182.1 | |||
XM_011515881.2 | XP_011514183.1 | |||
XM_011515882.2 | XP_011514184.1 | |||
AL832533.1 | 1 | 872 | ||
BC110606.1 | AAI10607.1 | |||
BC110607.1 | ||||
D83492.1 | BAA21560.1 | |||
EU054308.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv949965 | Chr.7:142862343 - 142878755 on Build GRCh38 | Deletion | TRPV6 EPHB6 |
nsv831168 | Chr.7:142830591 - 143025918 on Build GRCh38 | Gain | C7orf34 KEL TRPV6 TRPV5 EPHB6 |
esv2759569 | Chr.7:142434958 - 143027060 on Build GRCh38 | Loss | PRSS2 C7orf34 TRBJ2-2 TRPV6 TRBC1 OR9A2 TRBV19 MTRNR2L6 TRBV27 TRPV5 TRBV6-5 PRSS1 TRBV21-1 TRBV23-1 KEL TRBV16 TRBV5-4 EPHB6 |
nsv526015 | Chr.7:142864981 - 142944120 on Build GRCh38 | Gain | C7orf34 KEL TRPV6 TRPV5 EPHB6 |
nsv520735 | Chr.7:142865498 - 142944120 on Build GRCh38 | Loss | C7orf34 KEL TRPV6 TRPV5 EPHB6 |
nsv428187 | Chr.7:142618692 - 142909521 on Build GRCh38 | Loss | PRSS2 TRBJ2-2 PRSS1 TRBV21-1 TRBV23-1 TRPV6 TRBC1 TRBV19 MTRNR2L6 TRBV27 TRPV5 EPHB6 |
nsv1030700 | Chr.7:142867398 - 143010429 on Build GRCh38 | Gain | C7orf34 KEL TRPV6 TRPV5 EPHB6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |