Assay Details
Target Gene Details
Entrez Gene ID: | 10320 |
Gene Name: | IKAROS family zinc finger 1 |
Gene Aliases: |
CVID13, Hs.54452, IK1, IKAROS, LYF1, LyF-1, PPP1R92, PRO0758, ZNFN1A1 |
Location: |
Chr.7:50303453-50405101 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv950215 | Chr.7:50343503 - 50353202 on Build GRCh38 | Duplication | IKZF1 |
nsv606897 | Chr.7:49632287 - 50387038 on Build GRCh38 | Gain | VWC2 C7orf72 ZPBP IKZF1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |