Assay Details
Target Gene Details
Entrez Gene ID: | 101927811 |
Gene Name: | uncharacterized LOC101927811 |
Gene Aliases: |
- |
Location: |
Chr.7:20328301-20331762 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101927811 | NR_110119.1 | 4 | 443 | |
NR_110120.1 | 3 | 350 | ||
AV707592.1 | 4 | 375 | ||
AW291857.1 | ||||
HY150107.1 | 3 | 351 |