Assay Details
Target Gene Details
Entrez Gene ID: | 6239 |
Gene Name: | ras responsive element binding protein 1 |
Gene Aliases: |
FINB, HNT, LZ321, RREB-1, Zep-1 |
Location: |
Chr.6:7107597-7251980 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RREB1 | NM_001003698.3 | NP_001003698.1 | ||
NM_001003699.3 | NP_001003699.1 | |||
NM_001003700.1 | NP_001003700.1 | |||
NM_001168344.1 | NP_001161816.1 | |||
XM_011514794.2 | XP_011513096.1 | |||
XM_011514795.2 | XP_011513097.1 | |||
AB019254.1 | BAD32776.1 | |||
AB019351.1 | BAD32778.1 | |||
AB019352.1 | BAD32779.1 | |||
BI818966.1 | ||||
BP365591.1 | ||||
BX647723.1 | ||||
D49835.1 | BAA23165.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3890787 | Chr.6:6996928 - 7981438 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)