Assay Details
Target Gene Details
Entrez Gene ID: | 51567 |
Gene Name: | tyrosyl-DNA phosphodiesterase 2 |
Gene Aliases: |
AD022, EAP2, EAPII, TTRAP, dJ30M3.3, hTDP2 |
Location: |
Chr.6:24649977-24666887 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TDP2 | NM_016614.2 | NP_057698.2 | ||
AF201687.1 | AAG35600.1 | |||
AF223469.1 | AAF64144.1 | |||
AJ269473.1 | CAB92966.1 | |||
AJ420495.1 | ||||
AK002168.1 | BAA92119.1 | |||
AK296623.1 | ||||
AK298699.1 | ||||
AK308836.1 | ||||
BC017553.2 | AAH17553.1 | |||
BC110375.1 | AAI10376.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv601165 | Chr.6:24632414 - 24665779 on Build GRCh38 | Loss | TDP2 KIAA0319 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |