Assay Details
Target Gene Details
Entrez Gene ID: | 117247 |
Gene Name: | solute carrier family 16 member 10 |
Gene Aliases: |
MCT10, PRO0813, TAT1 |
Location: |
Chr.6:111087503-111227125 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC16A10 | NM_018593.4 | NP_061063.2 | ||
XM_006715329.2 | XP_006715392.1 | |||
XM_011535422.2 | XP_011533724.1 | |||
AB057445.1 | BAB84670.1 | |||
AK126183.1 | ||||
BC034031.1 | ||||
BC066985.1 | AAH66985.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv604517 | Chr.6:111031543 - 111259358 on Build GRCh38 | Gain | MFSD4B SLC16A10 GSTM2P1 |
nsv1020627 | Chr.6:110982539 - 111302511 on Build GRCh38 | Gain | MFSD4B RPF2 SLC16A10 GSTM2P1 REV3L |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |