Assay Details
Target Gene Details
Entrez Gene ID: | 81847 |
Gene Name: | ring finger protein 146 |
Gene Aliases: |
- |
Location: |
Chr.6:127266340-127289893 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |