Assay Details
Target Gene Details
Entrez Gene ID: | 55788 |
Gene Name: | LMBR1 domain containing 1 |
Gene Aliases: |
C6orf209, LMBD1, MAHCF, NESI |
Location: |
Chr.6:69675749-69797157 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LMBRD1 | NM_018368.3 | NP_060838.3 | ||
XM_011535941.1 | XP_011534243.1 | |||
AF113224.1 | AAG39295.1 | |||
AF208863.1 | AAF64277.1 | |||
AF211480.1 | AAK26247.1 | |||
AK002102.1 | BAA92087.1 | |||
AK225187.1 | ||||
AK225537.1 | ||||
AK290069.1 | ||||
AY136817.1 | AAN11301.1 | |||
BC010360.1 | AAH10360.1 | |||
BC047073.1 | AAH47073.1 | |||
BX647737.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |