Assay Details
Target Gene Details
Entrez Gene ID: | 2880 |
Gene Name: | glutathione peroxidase 5 |
Gene Aliases: |
HEL-S-75p |
Location: |
Chr.6:28526012-28534951 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPX5 | NM_001509.2 | NP_001500.1 | ||
NM_003996.3 | NP_003987.2 | |||
AJ005277.1 | CAA06463.1 | |||
BC128159.1 | ||||
BC128160.1 | ||||
FJ460514.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1074388 | Chr.6:28510222 - 29115423 on Build GRCh38 | Deletion | LOC100129636 OR2J3 OR2W1 LINC01623 ZNF311 HCG16 LINC00533 HCG14 TRIM27 LINC01556 ZBED9 GPX6 GPX5 OR2B3 |
dgv3296n106 | Chr.6:28510223 - 28623423 on Build GRCh38 | Deletion | ZBED9 GPX6 GPX5 |
nsv601211 | Chr.6:28515705 - 28553539 on Build GRCh38 | Loss | GPX6 GPX5 |
esv3576110 | Chr.6:28449711 - 28681348 on Build GRCh38 | Gain | ZBED9 GPX6 GPX5 LINC00533 |
nsv830621 | Chr.6:28506174 - 28707932 on Build GRCh38 | Gain | ZBED9 GPX6 GPX5 LINC00533 |
nsv601210 | Chr.6:28449445 - 28606638 on Build GRCh38 | Gain | ZBED9 GPX6 GPX5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |