Assay Details
Target Gene Details
Entrez Gene ID: | 57497 |
Gene Name: | leucine rich repeat and fibronectin type III domain containing 2 |
Gene Aliases: |
FIGLER2, KIAA1246, SALM1 |
Location: |
Chr.6:40391589-40587464 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRFN2 | NM_020737.2 | NP_065788.1 | ||
XM_011514761.2 | XP_011513063.1 | |||
XM_011514762.2 | XP_011513064.1 | |||
XM_017011110.1 | XP_016866599.1 | |||
AB033072.1 | ||||
AK291474.1 | ||||
BC142616.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv5273 | Chr.6:40373795 - 40418819 on Build GRCh38 | Deletion | TDRG1 LRFN2 |
esv2763989 | Chr.6:40376720 - 40464040 on Build GRCh38 | Loss | TDRG1 LRFN2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |