Assay Details
Target Gene Details
Entrez Gene ID: | 80144 |
Gene Name: | Fraser extracellular matrix complex subunit 1 |
Gene Aliases: |
- |
Location: |
Chr.4:78057323-78544269 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FRAS1 | NM_001166133.1 | NP_001159605.1 | ||
NM_025074.6 | NP_079350.5 | |||
XM_006714316.2 | XP_006714379.1 | |||
BC131820.1 | ||||
BC131821.1 | ||||
DA725113.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1009633 | Chr.4:77966419 - 78731082 on Build GRCh38 | Gain | ANXA3 LOC101928893 FRAS1 LINC01094 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |