Assay Details
Target Gene Details
Entrez Gene ID: | 10417 |
Gene Name: | spondin 2 |
Gene Aliases: |
DIL-1, DIL1, M-SPONDIN, MINDIN |
Location: |
Chr.4:1166932-1208962 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPON2 | NM_001199021.1 | NP_001185950.1 | ||
DA087369.1 | ||||
DA201089.1 | ||||
DB249869.1 | ||||
DB254983.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss | UVSSA PDE6B GAK MAEA DGKQ LOC107986211 ATP5I MFSD7 TMED11P PCGF3 CRIPAK FGFRL1 TMEM175 LOC101928548 SLC26A1 LOC105374344 IDUA NKX1-1 CTBP1 MYL5 LOC101928521 LOC100130872 RNF212 SPON2 CTBP1-AS CTBP1-AS2 CPLX1 LOC100129917 |
nsv593278 | Chr.4:1165892 - 1421910 on Build GRCh38 | Loss | UVSSA CTBP1-AS2 CRIPAK LOC100130872 MAEA LOC101928548 SPON2 CTBP1-AS NKX1-1 CTBP1 |
nsv950331 | Chr.4:1164113 - 1239212 on Build GRCh38 | Deletion | LOC100130872 LOC101928548 SPON2 CTBP1-AS CTBP1 |
nsv519106 | Chr.4:1165203 - 1181073 on Build GRCh38 | Loss | SPON2 |
nsv1012892 | Chr.4:1093083 - 1182842 on Build GRCh38 | Loss | TMED11P RNF212 SPON2 LOC105374344 |
nsv829835 | Chr.4:1103798 - 1310451 on Build GRCh38 | Gain | TMED11P CTBP1-AS2 LOC100130872 MAEA LOC101928548 RNF212 SPON2 LOC105374344 CTBP1-AS CTBP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |