Assay Details
Target Gene Details
Entrez Gene ID: | 56034 |
Gene Name: | platelet derived growth factor C |
Gene Aliases: |
FALLOTEIN, SCDGF |
Location: |
Chr.4:156761611-156971394 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDGFC | NM_016205.2 | NP_057289.1 | ||
NR_036641.1 | ||||
XM_011532124.2 | XP_011530426.1 | |||
XM_017008455.1 | XP_016863944.1 | |||
XM_017008456.1 | XP_016863945.1 | |||
AB033831.1 | BAB03266.1 | |||
AF091434.1 | AAF00049.1 | |||
AF244813.1 | AAF80597.1 | |||
AF260738.1 | AAK51637.1 | |||
AK300480.1 | ||||
AK304638.1 | ||||
AK313817.1 | ||||
AM922296.1 | ||||
AY358493.1 | AAQ88857.1 | |||
BC136662.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830122 | Chr.4:156695746 - 156892693 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)