Assay Details
Target Gene Details
Entrez Gene ID: | 27068 |
Gene Name: | pyrophosphatase (inorganic) 2 |
Gene Aliases: |
HSPC124, SID6-306 |
Location: |
Chr.4:105369077-105474070 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPA2 | NM_006903.4 | NP_008834.3 | ||
NM_176866.2 | NP_789842.2 | |||
NM_176867.3 | NP_789843.2 | |||
NM_176869.2 | NP_789845.1 | |||
AB026722.1 | BAA84701.1 | |||
AF161473.1 | AAF29088.1 | |||
AF217187.1 | AAG36781.1 | |||
AK000466.1 | BAA91184.1 | |||
AK289607.1 | ||||
AL136655.1 | CAB66590.2 | |||
BC022803.1 | AAH22803.1 | |||
BC039462.1 | AAH39462.2 | |||
BC057219.1 | AAH57219.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1000196 | Chr.4:103475625 - 106148054 on Build GRCh38 | Loss | LOC101929529 LOC101929468 INTS12 GSTCD NPNT TET2 ARHGEF38-IT1 LOC101929577 TBCK PPA2 TET2-AS1 CXXC4 TACR3 ARHGEF38 |
nsv830027 | Chr.4:105300853 - 105485581 on Build GRCh38 | Gain | TET2-AS1 PPA2 |
nsv595007 | Chr.4:104948391 - 105445052 on Build GRCh38 | Loss | TET2 TET2-AS1 PPA2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |