Assay Details
Target Gene Details
Entrez Gene ID: | 100505875 |
Gene Name: | long intergenic non-protein coding RNA 1088 |
Gene Aliases: |
- |
Location: |
Chr.4:78971748-79308799 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC01088 | NR_038342.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829979 | Chr.4:78898014 - 79098005 on Build GRCh38 | Gain | LINC01088 PAQR3 BMP2K |
esv2727859 | Chr.4:78716995 - 79557290 on Build GRCh38 | Deletion | LINC01088 NAA11 PAQR3 MIR5096 LINC00989 BMP2K GK2 LOC101928893 |
nsv1006026 | Chr.4:78111837 - 79966019 on Build GRCh38 | Loss | ANTXR2 FRAS1 PAQR3 MIR5096 BMP2K GK2 ANXA3 LINC01088 NAA11 LINC01094 PCAT4 LINC00989 LOC101928893 |