Assay Details
Target Gene Details
Entrez Gene ID: | 441024 |
Gene Name: | methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like |
Gene Aliases: |
- |
Location: |
Chr.4:74123280-74303099 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3569519 | Chr.4:74142954 - 74255926 on Build GRCh38 | Loss | MTHFD2L |
nsv1004768 | Chr.4:73908655 - 74243862 on Build GRCh38 | Gain | PPBP PF4 PPBPP2 CXCL3 CXCL2 MTHFD2L CXCL5 |
nsv525980 | Chr.4:73865809 - 74556070 on Build GRCh38 | Gain | PPBP PF4 EPGN LOC105377276 PPBPP2 CXCL3 CXCL2 AREG EREG MTHFD2L CXCL5 CXCL1 |
dgv5320n100 | Chr.4:74147724 - 74265726 on Build GRCh38 | Loss | MTHFD2L |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |