Assay Details
Target Gene Details
Entrez Gene ID: | 133121 |
Gene Name: | ectonucleotide pyrophosphatase/phosphodiesterase 6 |
Gene Aliases: |
NPP6 |
Location: |
Chr.4:184088706-184217961 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ENPP6 | NM_153343.3 | NP_699174.1 | ||
AK057370.1 | BAB71455.1 | |||
AY358676.1 | AAQ89039.1 | |||
BC035035.2 | AAH35035.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv521658 | Chr.4:184057338 - 184121636 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)