Assay Details
Target Gene Details
Entrez Gene ID: | 4886 |
Gene Name: | neuropeptide Y receptor Y1 |
Gene Aliases: |
NPY1-R, NPYR |
Location: |
Chr.4:163323962-163344847 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NPY1R | XM_005263031.3 | XP_005263088.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1015505 | Chr.4:161989825 - 163516137 on Build GRCh38 | Gain | NAF1 LOC101928052 TMA16 MIR4454 TKTL2 FSTL5 NPY5R NPY1R |
nsv1030184 | Chr.4:162402427 - 164455529 on Build GRCh38 | Loss | NAF1 ANP32C TMA16 MARCH1 MIR4454 TKTL2 NPY5R NPY1R |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |