Assay Details
Target Gene Details
Entrez Gene ID: | 55016 |
Gene Name: | membrane associated ring-CH-type finger 1 |
Gene Aliases: |
MARCH-I, RNF171 |
Location: |
Chr.4:163524298-164384050 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MARCH1 | NM_001166373.1 | NP_001159845.1 | ||
NM_017923.3 | NP_060393.1 | |||
XM_011532055.2 | XP_011530357.1 | |||
XM_017008334.1 | XP_016863823.1 | |||
XM_017008335.1 | XP_016863824.1 | |||
XM_017008336.1 | XP_016863825.1 | |||
AK000675.1 | BAA91319.1 | |||
AK289711.1 | ||||
AK301408.1 | ||||
AL713759.1 | CAD28529.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1030184 | Chr.4:162402427 - 164455529 on Build GRCh38 | Loss | NAF1 TKTL2 MARCH1 ANP32C TMA16 NPY5R MIR4454 NPY1R |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |