Assay Details
Target Gene Details
Entrez Gene ID: | 79625 |
Gene Name: | neuron derived neurotrophic factor |
Gene Aliases: |
C4orf31, NORD |
Location: |
Chr.4:121035627-121072518 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDNF | NM_024574.3 | NP_078850.3 | ||
AK289615.1 | ||||
AY358821.1 | AAQ89180.1 | |||
BC019351.1 | ||||
BX359842.2 | ||||
DA807168.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)