Assay Details
Target Gene Details
Entrez Gene ID: | 442117 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase-like 6 |
Gene Aliases: |
GALNACT20, GALNT17, GalNAc-T6L |
Location: |
Chr.4:171812690-173040408 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GALNTL6 | NM_001034845.2 | NP_001030017.2 | ||
XM_011531993.2 | XP_011530295.1 | |||
XM_017008243.1 | XP_016863732.1 | |||
XM_017008244.1 | XP_016863733.1 | |||
AJ626725.1 | CAF25036.1 | |||
AK295464.1 | ||||
BC047551.1 | AAH47551.1 | |||
GU220060.1 |
Target Gene Details
Entrez Gene ID: | 101928314 |
Gene Name: | uncharacterized LOC101928314 |
Gene Aliases: |
- |
Location: |
Chr.4:172629932-172726878 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101928314 | NR_125894.1 | |||
NR_125895.1 | ||||
BI461914.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv1042e214 | Chr.4:172635961 - 172745180 on Build GRCh38 | Gain | GALNTL6 LOC101928314 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |