Assay Details
Target Gene Details
Entrez Gene ID: | 10417 |
Gene Name: | spondin 2 |
Gene Aliases: |
DIL-1, DIL1, M-SPONDIN, MINDIN |
Location: |
Chr.4:1166932-1208962 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPON2 | NM_001199021.1 | NP_001185950.1 | ||
DA201089.1 | ||||
DB249869.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv8891n54 | Chr.4:1181073 - 1261142 on Build GRCh38 | Loss | SPON2 LOC100130872 LOC101928548 CTBP1-AS2 CTBP1 CTBP1-AS |
nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss | MFSD7 FGFRL1 TMEM175 NKX1-1 LOC100129917 RNF212 CTBP1-AS2 CTBP1 GAK SLC26A1 PCGF3 LOC101928548 MAEA IDUA LOC107986211 LOC101928521 ATP5I DGKQ LOC105374344 MYL5 SPON2 TMED11P CPLX1 LOC100130872 UVSSA CRIPAK PDE6B CTBP1-AS |
nsv593278 | Chr.4:1165892 - 1421910 on Build GRCh38 | Loss | SPON2 LOC100130872 LOC101928548 UVSSA CRIPAK MAEA NKX1-1 CTBP1-AS2 CTBP1 CTBP1-AS |
nsv950331 | Chr.4:1164113 - 1239212 on Build GRCh38 | Deletion | SPON2 LOC100130872 LOC101928548 CTBP1 CTBP1-AS |
nsv508988 | Chr.4:1190955 - 1358870 on Build GRCh38 | Insertion | SPON2 LOC100130872 LOC101928548 UVSSA MAEA CTBP1-AS2 CTBP1 CTBP1-AS |
nsv829835 | Chr.4:1103798 - 1310451 on Build GRCh38 | Gain | LOC105374344 SPON2 TMED11P LOC100130872 LOC101928548 MAEA RNF212 CTBP1-AS2 CTBP1 CTBP1-AS |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |