Assay Details
Target Gene Details
Entrez Gene ID: | 2560 |
Gene Name: | gamma-aminobutyric acid type A receptor beta1 subunit |
Gene Aliases: |
- |
Location: |
Chr.4:47031278-47438409 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GABRB1 | NM_000812.3 | NP_000803.2 | ||
XM_017007986.1 | XP_016863475.1 | |||
AK225167.1 | ||||
AK225686.1 | ||||
AK293691.1 | ||||
AK312720.1 | ||||
BC022449.1 | AAH22449.1 | |||
KJ534841.1 | ||||
KJ535054.1 | ||||
X14767.1 | CAA32875.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)