Assay Details
Target Gene Details
Entrez Gene ID: | 7358 |
Gene Name: | UDP-glucose 6-dehydrogenase |
Gene Aliases: |
GDH, UDP-GlcDH, UDPGDH, UGD |
Location: |
Chr.4:39498755-39527598 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UGDH | NM_001184700.1 | NP_001171629.1 | ||
NM_001184701.1 | NP_001171630.1 | |||
NM_003359.3 | NP_003350.1 | |||
XM_005262667.2 | XP_005262724.1 | |||
AF061016.1 | AAC36095.1 | |||
AJ007702.1 | CAA07609.1 | |||
AK097930.1 | ||||
AK297737.1 | ||||
AL708740.1 | ||||
AU131705.1 | ||||
BC022781.1 | AAH22781.1 | |||
CN405740.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1010784 | Chr.4:39437476 - 39571758 on Build GRCh38 | Gain | KLB RPL9 SMIM14 UGDH LOC401127 UGDH-AS1 LIAS |
nsv1010189 | Chr.4:39423149 - 39523510 on Build GRCh38 | Gain | KLB RPL9 UGDH LOC401127 LIAS |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |