Assay Details
Target Gene Details
Entrez Gene ID: | 200879 |
Gene Name: | lipase H |
Gene Aliases: |
AH, ARWH2, HYPT7, LAH2, LPDLR, PLA1B, mPA-PLA1 |
Location: |
Chr.3:185506262-185552661 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPH | NM_139248.2 | NP_640341.1 | ||
XM_006713529.3 | XP_006713592.1 | |||
XM_011512530.2 | XP_011510832.1 | |||
XM_011512531.2 | XP_011510833.1 | |||
XM_017005852.1 | XP_016861341.1 | |||
AK074229.1 | BAB85023.1 | |||
AK122651.1 | ||||
AK298380.1 | ||||
AY036912.1 | AAK63178.1 | |||
AY093498.1 | AAM18803.1 | |||
BC064941.1 | AAH64941.1 | |||
EF186228.1 | ||||
EF186229.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829818 | Chr.3:185411745 - 185567640 on Build GRCh38 | Loss | TMEM41A MAP3K13 LIPH |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |