Assay Details
Target Gene Details
Entrez Gene ID: | 686 |
Gene Name: | biotinidase |
Gene Aliases: |
- |
Location: |
Chr.3:15601352-15653709 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BTD | NM_001281726.1 | NP_001268655.1 | ||
XM_006713314.3 | XP_006713377.1 | |||
XM_011534041.2 | XP_011532343.1 | |||
XM_017007088.1 | XP_016862577.1 | |||
AI391654.1 | ||||
AK294301.1 | ||||
AK297033.1 | ||||
AK301838.1 | ||||
AK313252.1 | ||||
BC012099.1 | AAH12099.1 | |||
BE814688.1 | ||||
BM839350.1 | ||||
BM919775.1 | ||||
BP326502.1 | ||||
U03274.1 | AAC04318.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv979807 | Chr.3:15636527 - 15641078 on Build GRCh38 | Duplication | BTD |
More Information
Additional Information:
For this assay, SNP(s) [rs114461561] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |