Assay Details
Target Gene Details
Entrez Gene ID: | 4968 |
Gene Name: | 8-oxoguanine DNA glycosylase |
Gene Aliases: |
HMMH, HOGG1, MUTM, OGH1 |
Location: |
Chr.3:9749944-9788246 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OGG1 | XM_011533760.1 | XP_011532062.1 | ||
XM_017006494.1 | XP_016861983.1 | |||
XM_017006496.1 | XP_016861985.1 | |||
XM_017006497.1 | XP_016861986.1 | |||
XM_017006499.1 | XP_016861988.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1010768 | Chr.3:9502870 - 9873902 on Build GRCh38 | Gain | CIDEC ARPC4-TTLL3 TADA3 LOC107986062 CAMK1 MTMR14 ARPC4 RPUSD3 TTLL3 LHFPL4 CPNE9 BRPF1 OGG1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |