Assay Details
Target Gene Details
Entrez Gene ID: | 440944 |
Gene Name: | THUMPD3 antisense RNA 1 |
Gene Aliases: |
SETD5-AS1 |
Location: |
Chr.3:9385264-9397494 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
THUMPD3-AS1 | NR_027007.3 | 3 | 808 | |
NR_132780.1 | 4 | 882 | ||
NR_132781.1 | ||||
AI248388.1 | ||||
AK093640.1 | ||||
AL110136.1 | 3 | 804 | ||
AW237468.1 | ||||
DB567546.1 |