Assay Details
Target Gene Details
Entrez Gene ID: | 79443 |
Gene Name: | FYVE and coiled-coil domain containing 1 |
Gene Aliases: |
CATC2, CTRCT18, RUFY3, ZFYVE7 |
Location: |
Chr.3:45917899-45995824 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FYCO1 | NM_024513.3 | NP_078789.2 | ||
XM_006713333.3 | XP_006713396.1 | |||
XM_006713334.3 | XP_006713397.1 | |||
XM_011534111.2 | XP_011532413.1 | |||
AJ292348.2 | CAC33883.1 | |||
AK023397.1 | BAB14559.1 | |||
AK074165.1 | BAB84991.1 | |||
AL832358.1 | CAD91151.1 | |||
AL833308.2 | CAD89924.1 | |||
BC007218.1 | AAH07218.1 | |||
BC101468.1 | AAI01469.1 | |||
BC101470.1 | AAI01471.1 | |||
BC143368.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834678 | Chr.3:45762802 - 45968764 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)