Assay Details
Target Gene Details
Entrez Gene ID: | 1238 |
Gene Name: | atypical chemokine receptor 2 |
Gene Aliases: |
CCBP2, CCR10, CCR9, CMKBR9, D6, hD6 |
Location: |
Chr.3:42809472-42867283 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACKR2 | NM_001296.4 | NP_001287.2 | ||
AK313561.1 | ||||
BC008816.2 | AAH08816.1 | |||
BC011631.2 | AAH11631.1 | |||
BC020558.1 | AAH20558.1 | |||
BE901130.1 | ||||
DA853159.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |