Assay Details
Target Gene Details
Entrez Gene ID: | 5449 |
Gene Name: | POU class 1 homeobox 1 |
Gene Aliases: |
CPHD1, GHF-1, PIT1, POU1F1a, Pit-1 |
Location: |
Chr.3:87259633-87276587 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POU1F1 | NM_000306.3 | NP_000297.1 | ||
NM_001122757.2 | NP_001116229.1 | |||
D10216.1 | BAA01068.1 | |||
L18781.1 | AAA60093.1 | |||
X62429.1 | CAA44295.1 | |||
X72215.1 | CAA51017.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3596885 | Chr.3:87265407 - 87268233 on Build GRCh38 | Loss | POU1F1 |
esv3568885 | Chr.3:87182571 - 87518398 on Build GRCh38 | Loss | CHMP2B POU1F1 MIR4795 |
nsv527279 | Chr.3:86796784 - 87814130 on Build GRCh38 | Gain | CHMP2B LINC00506 POU1F1 VGLL3 HTR1F MIR4795 |
nsv470749 | Chr.3:87258397 - 87302230 on Build GRCh38 | Loss | POU1F1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |