Assay Details
Target Gene Details
Entrez Gene ID: | 23429 |
Gene Name: | RING1 and YY1 binding protein |
Gene Aliases: |
AAP1, APAP-1, DEDAF, YEAF1 |
Location: |
Chr.3:72374593-72446623 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RYBP | NM_012234.6 | NP_036366.3 | ||
AB029551.1 | BAA89486.1 | |||
AF179286.1 | AAD51858.1 | |||
AF227959.1 | AAK63197.1 | |||
AY228125.1 | AAO73587.1 | |||
BC014959.1 | AAH14959.1 | |||
BC036459.1 | AAH36459.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv954886 | Chr.3:72374650 - 72446149 on Build GRCh38 | Duplication | RYBP |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |