Assay Details
Target Gene Details
Entrez Gene ID: | 101928583 |
Gene Name: | uncharacterized LOC101928583 |
Gene Aliases: |
- |
Location: |
Chr.3:170467285-170755205 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101928583 | NR_135555.1 | |||
NR_135556.1 | ||||
NR_135557.1 | ||||
HY157714.1 |
Target Gene Details
Entrez Gene ID: | 57709 |
Gene Name: | solute carrier family 7 member 14 |
Gene Aliases: |
PPP1R142 |
Location: |
Chr.3:170459554-170586074 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC7A14 | NM_020949.2 | NP_066000.2 | ||
AB046833.1 | BAB13439.1 | |||
AK122655.1 | ||||
BC022968.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)