Assay Details
Target Gene Details
Entrez Gene ID: | 9626 |
Gene Name: | guanylate cyclase activator 1C |
Gene Aliases: |
GCAP3 |
Location: |
Chr.3:108907644-108955195 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GUCA1C | NM_005459.3 | NP_005450.3 | ||
XM_005247910.3 | XP_005247967.1 | |||
XM_011513334.2 | XP_011511636.1 | |||
AB593147.1 | ||||
AF110002.1 | AAD19944.1 | |||
AF110003.1 | AAD19945.1 | |||
AV732101.1 | ||||
BC103993.1 | AAI03994.1 | |||
BC103994.1 | AAI03995.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829662 | Chr.3:108821681 - 109005440 on Build GRCh38 | Gain | MORC1 TRAT1 GUCA1C |
nsv829663 | Chr.3:108916545 - 109070871 on Build GRCh38 | Gain | MORC1 GUCA1C |
dgv4836n100 | Chr.3:108877854 - 109296324 on Build GRCh38 | Gain | FLJ22763 MORC1 LINC00488 DPPA2 GUCA1C |
nsv522119 | Chr.3:108756961 - 109199392 on Build GRCh38 | Gain | FLJ22763 MORC1 LINC00488 TRAT1 RETNLB GUCA1C |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |