Assay Details
Target Gene Details
Entrez Gene ID: | 55773 |
Gene Name: | TBC1 domain family member 23 |
Gene Aliases: |
NS4ATP1 |
Location: |
Chr.3:100260817-100325252 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TBC1D23 | NM_001199198.2 | NP_001186127.1 | ||
NM_018309.4 | NP_060779.2 | |||
XM_017006840.1 | XP_016862329.1 | |||
XM_017006841.1 | XP_016862330.1 | |||
AB449910.1 | ||||
AK001908.1 | BAA91973.1 | |||
BC020955.1 | AAH20955.1 | |||
BX648032.1 | ||||
DA801258.1 | ||||
DB458401.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)