Assay Details
Target Gene Details
Entrez Gene ID: | 54859 |
Gene Name: | elongator acetyltransferase complex subunit 6 |
Gene Aliases: |
C3orf75, TMEM103 |
Location: |
Chr.3:47495640-47513711 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ELP6 | NM_001031703.2 | NP_001026873.2 | ||
XM_005265241.3 | XP_005265298.1 | |||
XM_011533851.2 | XP_011532153.1 | |||
XM_011533852.2 | XP_011532154.1 | |||
XM_011533853.2 | XP_011532155.1 | |||
XM_011533854.2 | XP_011532156.1 | |||
XM_011533856.2 | XP_011532158.1 | |||
XM_011533857.2 | XP_011532159.1 | |||
XM_017006669.1 | XP_016862158.1 | |||
XM_017006670.1 | XP_016862159.1 | |||
AK000218.1 | BAA91017.1 | |||
AK225721.1 | ||||
AK298202.1 | ||||
BC000623.2 | AAH00623.1 | |||
BC014204.2 | AAH14204.1 | |||
DQ787088.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3596083 | Chr.3:47485559 - 47529809 on Build GRCh38 | Gain | ELP6 |
nsv428417 | Chr.3:46638962 - 47778557 on Build GRCh38 | Loss | NRADDP PRSS46 MYL3 TMIE CSPG5 KLHL18 PRSS50 ALS2CL NBEAL2 PRSS43 SCAP SNORD13P3 PTH1R PRSS42 KIF9-AS1 SMARCC1 PTPN23 ELP6 CCDC12 PRSS45 KIF9 SETD2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |