Assay Details
Target Gene Details
Entrez Gene ID: | 10803 |
Gene Name: | C-C motif chemokine receptor 9 |
Gene Aliases: |
CC-CKR-9, CDw199, GPR-9-6, GPR28 |
Location: |
Chr.3:45886504-45903177 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCR9 | NM_001256369.1 | NP_001243298.1 | ||
NM_006641.3 | NP_006632.2 | |||
NM_031200.2 | NP_112477.1 | |||
AF145207.1 | AAD38539.1 | |||
AF145439.1 | AAF66699.1 | |||
AF145440.1 | AAF66700.1 | |||
AJ132337.1 | CAB43477.1 | |||
AK312824.1 | ||||
AK313366.1 | ||||
BC069678.1 | AAH69678.1 | |||
DB103756.1 | ||||
DB132476.1 |
Target Gene Details
Entrez Gene ID: | 54585 |
Gene Name: | leucine zipper transcription factor like 1 |
Gene Aliases: |
BBS17 |
Location: |
Chr.3:45823316-45916042 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LZTFL1 | NM_001276378.1 | NP_001263307.1 | ||
NM_001276379.1 | NP_001263308.1 | |||
XM_017006644.1 | XP_016862133.1 | |||
XM_017006645.1 | XP_016862134.1 | |||
AK093705.1 | ||||
AK303416.1 | ||||
DC410047.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834678 | Chr.3:45762802 - 45968764 on Build GRCh38 | Gain | CCR9 SLC6A20 LZTFL1 CXCR6 FYCO1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |