Assay Details
Target Gene Details
Entrez Gene ID: | 79750 |
Gene Name: | zinc finger protein 385D |
Gene Aliases: |
ZNF659 |
Location: |
Chr.3:21418419-22372641 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF385D | NM_024697.2 | NP_078973.1 | ||
XM_011534122.1 | XP_011532424.1 | |||
XM_017007191.1 | XP_016862680.1 | |||
XM_017007192.1 | XP_016862681.1 | |||
XM_017007193.1 | XP_016862682.1 | |||
XM_017007194.1 | XP_016862683.1 | |||
XM_017007195.1 | XP_016862684.1 | |||
XM_017007196.1 | XP_016862685.1 | |||
XM_017007197.1 | XP_016862686.1 | |||
XM_017007198.1 | XP_016862687.1 | |||
XM_017007199.1 | XP_016862688.1 | |||
XM_017007200.1 | XP_016862689.1 | |||
XM_017007201.1 | XP_016862690.1 | |||
XM_017007202.1 | XP_016862691.1 | |||
AK026072.1 | BAB15350.1 | |||
AK310298.1 | ||||
BC007212.1 | AAH07212.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1006471 | Chr.3:21344135 - 21555463 on Build GRCh38 | Gain | ZNF385D ZNF385D-AS1 VENTXP7 |
nsv834630 | Chr.3:21270127 - 21449835 on Build GRCh38 | Loss | ZNF385D VENTXP7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |