Assay Details
Target Gene Details
Entrez Gene ID: | 100126793 |
Gene Name: | ghrelin opposite strand/antisense RNA |
Gene Aliases: |
GHRL-AS1, GHRLAS, NCRNA00068 |
Location: |
Chr.3:10280952-10293449 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GHRLOS | NR_004431.3 | |||
NR_024144.2 | ||||
NR_024145.2 | ||||
NR_073566.1 | ||||
NR_073567.1 | ||||
NR_073568.1 | ||||
EU789528.1 | ||||
EU789529.1 | ||||
EU789530.1 | ||||
EU789534.1 | ||||
EU789535.1 | ||||
EU789536.1 | ||||
EU789537.1 | ||||
EU789538.1 | ||||
EU789539.1 | ||||
EU789540.1 | ||||
EU789541.1 | ||||
EU789542.1 | ||||
EU789543.1 | ||||
EU789544.1 | ||||
EU789545.1 | ||||
EU789546.1 | ||||
EU789547.1 | ||||
EU789548.1 | ||||
EU789549.1 | ||||
EU789550.1 | ||||
EU789551.1 | ||||
EU789552.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv436362 | Chr.3:10048344 - 11860564 on Build GRCh38 | Deletion | ATP2B2-IT2 TATDN2 ATP2B2 LOC101927467 ATG7 FANCD2 SLC6A11 GHRLOS LINC00852 IRAK2 HRH1 SLC6A1-AS1 MIR378B VHL BRK1 GHRL VGLL4 SEC13 MIR885 SLC6A1 TAMM41 FANCD2OS LINC00606 |
nsv508903 | Chr.3:10275353 - 10401894 on Build GRCh38 | Insertion | TATDN2 ATP2B2 GHRLOS GHRL LINC00852 SEC13 MIR885 MIR378B |
nsv589650 | Chr.3:10260488 - 10294102 on Build GRCh38 | Loss | TATDN2 GHRLOS GHRL LINC00852 |
dgv762e201 | Chr.3:10008967 - 11913136 on Build GRCh38 | Deletion | ATP2B2-IT2 LOC101927467 LOC401052 SLC6A11 FANCD2P2 EMC3 LINC00852 SLC6A1-AS1 BRK1 GHRL VGLL4 MIR885 TAMM41 FANCD2OS LINC00606 CIDECP TATDN2 ATP2B2 ATG7 FANCD2 GHRLOS IRAK2 HRH1 MIR378B VHL SEC13 SLC6A1 |
nsv1121603 | Chr.3:10071433 - 11889279 on Build GRCh38 | Deletion | ATP2B2-IT2 TATDN2 ATP2B2 LOC101927467 ATG7 FANCD2 SLC6A11 FANCD2P2 GHRLOS LINC00852 IRAK2 HRH1 SLC6A1-AS1 MIR378B VHL BRK1 GHRL VGLL4 SEC13 MIR885 SLC6A1 TAMM41 FANCD2OS LINC00606 |
nsv436869 | Chr.3:10064456 - 11877985 on Build GRCh38 | Insertion | ATP2B2-IT2 TATDN2 ATP2B2 LOC101927467 ATG7 FANCD2 SLC6A11 FANCD2P2 GHRLOS LINC00852 IRAK2 HRH1 SLC6A1-AS1 MIR378B VHL BRK1 GHRL VGLL4 SEC13 MIR885 SLC6A1 TAMM41 FANCD2OS LINC00606 |
nsv1073204 | Chr.3:9996548 - 11875005 on Build GRCh38 | Deletion | ATP2B2-IT2 LOC101927467 LOC401052 SLC6A11 FANCD2P2 EMC3 LINC00852 SLC6A1-AS1 EMC3-AS1 BRK1 GHRL VGLL4 MIR885 TAMM41 FANCD2OS LINC00606 CIDECP TATDN2 ATP2B2 ATG7 FANCD2 GHRLOS IRAK2 HRH1 MIR378B VHL SEC13 SLC6A1 |