Assay Details
Target Gene Details
Entrez Gene ID: | 100507582 |
Gene Name: | BHLHE40 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:4896809-4979961 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BHLHE40-AS1 | NR_037903.3 | |||
NR_125915.1 | ||||
NR_125916.1 | ||||
AK056892.1 | ||||
AK311646.1 | ||||
DA948054.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1005718 | Chr.3:4722315 - 5046200 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv470010 | Chr.3:4824636 - 4928648 on Build GRCh38 | Gain |
![]() ![]() |
nsv834606 | Chr.3:4835019 - 5057247 on Build GRCh38 | Loss |
![]() ![]() ![]() |
esv3893643 | Chr.3:4529987 - 4974421 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv589457 | Chr.3:4827274 - 4928648 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)