Assay Details
Target Gene Details
Entrez Gene ID: | 55825 |
Gene Name: | peroxisomal trans-2-enoyl-CoA reductase |
Gene Aliases: |
DCRRP, HPDHASE, HSA250303, PVIARL, SDR29C1, TERP |
Location: |
Chr.2:216029088-216081823 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PECR | AK296208.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1011506 | Chr.2:215969945 - 216310768 on Build GRCh38 | Gain | PKI55 TMEM169 MARCH4 PECR MREG XRCC5 |
nsv998837 | Chr.2:215982428 - 216143671 on Build GRCh38 | Gain | TMEM169 PECR MREG XRCC5 |