Assay Details
Target Gene Details
Entrez Gene ID: | 10152 |
Gene Name: | abl-interactor 2 |
Gene Aliases: |
ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB |
Location: |
Chr.2:203327874-203432173 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1001799 | Chr.2:202908732 - 205615330 on Build GRCh38 | Gain | PARD3B RPL12P16 RAPH1 CARF NBEAL1 ICOS WDR12 ABI2 CYP20A1 CTLA4 CD28 |
nsv3115 | Chr.2:203315491 - 203357486 on Build GRCh38 | Deletion | ABI2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |