Assay Details
Target Gene Details
Entrez Gene ID: | 151188 |
Gene Name: | ADP ribosylation factor like GTPase 6 interacting protein 6 |
Gene Aliases: |
AIP-6, AIP6, PFAAP1 |
Location: |
Chr.2:152717893-152761253 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARL6IP6 | NM_152522.5 | NP_689735.1 | ||
NR_024526.2 | ||||
XM_006712326.3 | XP_006712389.1 | |||
AF530059.1 | AAQ09943.1 | |||
AK023109.1 | ||||
AK094696.1 | ||||
AK315657.1 | ||||
AW967583.1 | ||||
BC028741.2 | AAH28741.1 | |||
BC070140.1 | AAH70140.1 | |||
BP200063.1 | ||||
BX437990.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834421 | Chr.2:152613595 - 152807721 on Build GRCh38 | Loss | PRPF40A FMNL2 ARL6IP6 |
esv34659 | Chr.2:150858366 - 152860232 on Build GRCh38 | Loss | NMI NEB MIR4773-2 ARL5A STAM2 PRPF40A LOC101929319 ARL6IP6 RBM43 TNFAIP6 CACNB4 MIR4773-1 FMNL2 RIF1 |
esv3592922 | Chr.2:151571476 - 153829714 on Build GRCh38 | Loss | RPRM NEB ARL5A STAM2 CACNB4 PRPF40A FMNL2 ARL6IP6 |
nsv834422 | Chr.2:152732934 - 152886555 on Build GRCh38 | Gain | ARL6IP6 |